Assay Design



Gene Transcripts

Gene Symbol
MANEAL
Entrez Gene ID
526978
Gene Name
mannosidase endo-alpha like
Gene Aliases
-
Chromosome Location
Chr.3: 108112880 - 108120963 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102105.1 NP_001095575.1 3 - 4 817 72
GenBank mRNA BC151266.1 - 3 - 4 817 72

View Full Product Details