Assay Design



Gene Transcripts

Gene Symbol
DRC3
Entrez Gene ID
527539
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
LRRC48
Chromosome Location
Chr.19: 34536472 - 34556492 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102111.1 NP_001095581.1 13 - 14 1888 114
GenBank mRNA BC149900.1 - 13 - 14 1888 114

View Full Product Details