Assay Design



Gene Transcripts

Gene Symbol
PIK3CG
Entrez Gene ID
530001
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
Gene Aliases
-
Chromosome Location
Chr.4: 47988347 - 48028141 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102125.2 NP_001095595.1 3 - 4 2177 89
GenBank mRNA BC149468.1 - 3 - 4 2178 89

View Full Product Details