Assay Design



Gene Transcripts

Gene Symbol
SLC19A3
Entrez Gene ID
537280
Gene Name
solute carrier family 19 member 3
Gene Aliases
-
Chromosome Location
Chr.2: 115812576 - 115843935 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102198.1 NP_001095668.1 2 - 3 304 119
GenBank mRNA BC151720.1 - 2 - 3 304 119

View Full Product Details