Assay Design



Gene Transcripts

Gene Symbol
LRRC14
Entrez Gene ID
539952
Gene Name
leucine rich repeat containing 14
Gene Aliases
-
Chromosome Location
Chr.14: 424286 - 428158 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102379.1 NP_001095849.1 3 - 4 1087 89
GenBank mRNA BC142136.1 - 3 - 4 1087 89

View Full Product Details