Assay Design



Gene Transcripts

Gene Symbol
PLA2G2F
Entrez Gene ID
541294
Gene Name
phospholipase A2 group IIF
Gene Aliases
-
Chromosome Location
Chr.2: 132496553 - 132509449 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102522.1 NP_001095992.1 3 - 4 423 71
GenBank mRNA BC147912.1 - 3 - 4 423 71

View Full Product Details