Assay Design



Gene Transcripts

Gene Symbol
MAFF
Entrez Gene ID
617914
Gene Name
MAF bZIP transcription factor F
Gene Aliases
-
Chromosome Location
Chr.5: 109963645 - 109972831 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103300.1 NP_001096770.1 2 - 3 271 75
GenBank mRNA BC151584.1 - 2 - 3 271 75

View Full Product Details