Assay Design



Gene Transcripts

Gene Symbol
SLC46A3
Entrez Gene ID
781117
Gene Name
solute carrier family 46 member 3
Gene Aliases
-
Chromosome Location
Chr.12: 31440779 - 31458812 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103303.2 NP_001096773.1 2 - 3 613 127
GenBank mRNA BC140697.1 - 2 - 3 614 127

View Full Product Details