Assay Design



Gene Transcripts

Gene Symbol
IRX3
Entrez Gene ID
540417
Gene Name
iroquois homeobox 3
Gene Aliases
-
Chromosome Location
Chr.18: 22615021 - 22618410 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104996.2 NP_001098466.1 3 - 4 1615 85
GenBank mRNA BC133453.1 - 3 - 4 1616 85

View Full Product Details