Assay Design



Gene Transcripts

Gene Symbol
EFNA2
Entrez Gene ID
614453
Gene Name
ephrin A2
Gene Aliases
-
Chromosome Location
Chr.7: 43722742 - 43737301 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105000.1 NP_001098470.1 3 - 4 552 85
GenBank mRNA BC140584.1 - 3 - 4 552 85

View Full Product Details