Assay Design



Gene Transcripts

Gene Symbol
GALR2
Entrez Gene ID
618629
Gene Name
galanin receptor 2
Gene Aliases
-
Chromosome Location
Chr.19: 55583908 - 55586715 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105010.1 NP_001098480.1 1 - 2 683 75
GenBank mRNA BC147995.1 - 1 - 2 683 75

View Full Product Details