Assay Design



Gene Transcripts

Gene Symbol
CNST
Entrez Gene ID
541076
Gene Name
consortin, connexin sorting protein
Gene Aliases
-
Chromosome Location
Chr.16: 30698935 - 30802211 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105256.3 NP_001098726.1 5 - 6 815 127

View Full Product Details