Assay Design



Gene Transcripts

Gene Symbol
INO80C
Entrez Gene ID
533426
Gene Name
INO80 complex subunit C
Gene Aliases
-
Chromosome Location
Chr.24: 21583219 - 21608154 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105404.1 NP_001098874.1 1 - 2 162 62
GenBank mRNA BC134592.1 - 1 - 2 162 62

View Full Product Details