Assay Design



Gene Transcripts

Gene Symbol
RNASE13
Entrez Gene ID
539741
Gene Name
ribonuclease A family member 13 (inactive)
Gene Aliases
RAL1
Chromosome Location
Chr.10: 26021932 - 26071803 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105426.1 NP_001098896.1 1 - 2 71 68
GenBank mRNA BC134656.1 - 1 - 2 71 68

View Full Product Details