Assay Design



Gene Transcripts

Gene Symbol
SYPL1
Entrez Gene ID
615728
Gene Name
synaptophysin like 1
Gene Aliases
-
Chromosome Location
Chr.4: 47150517 - 47180246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110088.1 NP_001103558.1 2 - 3 140 66
GenBank mRNA BC112703.1 - 2 - 3 140 66

View Full Product Details