Assay Design



Gene Transcripts

Gene Symbol
LDHC
Entrez Gene ID
537256
Gene Name
lactate dehydrogenase C
Gene Aliases
-
Chromosome Location
Chr.29: 26253200 - 26286422 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113249.1 NP_001106720.1 7 - 8 1123 85
GenBank mRNA DT837693.1 - 5 - 6 672 85

View Full Product Details