Assay Design



Gene Transcripts

Gene Symbol
PRPS2
Entrez Gene ID
537688
Gene Name
phosphoribosyl pyrophosphate synthetase 2
Gene Aliases
-
Chromosome Location
Chr.X: 130797766 - 130827907 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114152.1 NP_001107624.1 4 - 5 412 78

View Full Product Details