Assay Design



Gene Transcripts

Gene Symbol
TFB1M
Entrez Gene ID
533438
Gene Name
transcription factor B1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.9: 91865417 - 91932613 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114610.1 NP_001108082.1 6 - 7 844 87
GenBank mRNA DT811342.1 - 6 - 7 837 87

View Full Product Details