Assay Design



Gene Transcripts

Gene Symbol
CMTM3
Entrez Gene ID
787512
Gene Name
CKLF like MARVEL transmembrane domain containing 3
Gene Aliases
-
Chromosome Location
Chr.18: 34347394 - 34357748 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099399.1 NP_001092869.1 3 - 4 443 71
GenBank mRNA BC140678.1 - 3 - 4 443 71

View Full Product Details