Assay Design



Gene Transcripts

Gene Symbol
MOB2
Entrez Gene ID
789811
Gene Name
MOB kinase activator 2
Gene Aliases
HCCA2
Chromosome Location
Chr.29: 49990596 - 50043290 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099732.1 NP_001093202.1 2 - 3 262 78
GenBank mRNA BC142101.1 - 2 - 3 262 78

View Full Product Details