Assay Design



Gene Transcripts

Gene Symbol
FIZ1
Entrez Gene ID
789631
Gene Name
FLT3 interacting zinc finger 1
Gene Aliases
-
Chromosome Location
Chr.18: 61942342 - 61947503 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101308.1 NP_001094778.1 3 - 4 809 153
GenBank mRNA BC149069.1 - 3 - 4 809 153

View Full Product Details