Assay Design



Gene Transcripts

Gene Symbol
ZFP30
Entrez Gene ID
100124429
Gene Name
ZFP30 zinc finger protein
Gene Aliases
-
Chromosome Location
Chr.18: 47613683 - 47637327 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102539.1 NP_001096009.1 3 - 4 326 83
GenBank mRNA BC151289.1 - 3 - 4 326 83

View Full Product Details