Assay Design



Gene Transcripts

Gene Symbol
KHNYN
Entrez Gene ID
100124517
Gene Name
KH and NYN domain containing
Gene Aliases
-
Chromosome Location
Chr.10: 20734751 - 20744735 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102551.1 NP_001096021.1 2 - 3 344 90
GenBank mRNA BC151400.1 - 2 - 3 344 90

View Full Product Details