Assay Design



Gene Transcripts

Gene Symbol
CRTAM
Entrez Gene ID
781864
Gene Name
cytotoxic and regulatory T cell molecule
Gene Aliases
-
Chromosome Location
Chr.15: 33457290 - 33486131 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105457.1 NP_001098927.1 3 - 4 388 80
GenBank mRNA BC134770.1 - 3 - 4 388 80

View Full Product Details