Assay Design



Gene Transcripts

Gene Symbol
WDR82
Entrez Gene ID
783336
Gene Name
WD repeat domain 82
Gene Aliases
WDR82P1
Chromosome Location
Chr.22: 48629513 - 48648119 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105459.1 NP_001098929.1 1 - 2 179 76
GenBank mRNA BC153287.1 - 1 - 2 179 76

View Full Product Details