Assay Design



Gene Transcripts

Gene Symbol
FLACC1
Entrez Gene ID
784225
Gene Name
flagellum associated containing coiled-coil domains 1
Gene Aliases
ALS2CR12
Chromosome Location
Chr.2: 89886177 - 89924890 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105461.1 NP_001098931.1 7 - 8 942 61
GenBank mRNA BC123837.1 - 7 - 8 942 61

View Full Product Details