Assay Design



Gene Transcripts

Gene Symbol
SNTB2
Entrez Gene ID
786534
Gene Name
syntrophin beta 2
Gene Aliases
-
Chromosome Location
Chr.18: 36384453 - 36448425 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105466.1 NP_001098936.1 3 - 4 1033 69
GenBank mRNA BC134712.1 - 3 - 4 1033 69

View Full Product Details