Assay Design



Gene Transcripts

Gene Symbol
PCYOX1
Entrez Gene ID
100125835
Gene Name
prenylcysteine oxidase 1
Gene Aliases
-
Chromosome Location
Chr.11: 68555330 - 68599796 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105474.2 NP_001098944.1 4 - 5 739 70
GenBank mRNA BC149440.1 - 4 - 5 740 70

View Full Product Details