Assay Design



Gene Transcripts

Gene Symbol
RAB8A
Entrez Gene ID
100125881
Gene Name
RAB8A, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.7: 6874793 - 6894924 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105481.1 NP_001098951.1 5 - 6 424 90
GenBank mRNA BC123755.1 - 5 - 6 424 90

View Full Product Details