Assay Design



Gene Transcripts

Gene Symbol
PCDHGC3
Entrez Gene ID
100125301
Gene Name
protocadherin gamma subfamily C, 3
Gene Aliases
-
Chromosome Location
Chr.7: 52565851 - 52598880 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114080.1 NP_001107552.1 1 - 2 2563 69
GenBank mRNA BC140535.1 - 1 - 2 2563 69

View Full Product Details