Assay Design
Gene Transcripts
- Gene Symbol
- PAG5
- Entrez Gene ID
-
337899
- Gene Name
- pregnancy-associated glycoprotein 5
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 37788685 - 37797590 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176616.2 |
NP_788789.1 |
3 - 4 |
344 |
145 |
GenBank mRNA |
AF020507.1 |
- |
3 - 4 |
344 |
145 |
View Full Product Details