Assay Design
Gene Transcripts
- Gene Symbol
- PAG19
- Entrez Gene ID
-
337913
- Gene Name
- pregnancy-associated glycoprotein 19
- Gene Aliases
-
-
- Chromosome Location
-
Chr.29: 38684887 - 38693885 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_176628.1 |
NP_788801.1 |
7 - 8 |
888 |
165 |
GenBank mRNA |
AF192336.1 |
- |
7 - 8 |
888 |
165 |
View Full Product Details