Assay Design



Gene Transcripts

Gene Symbol
LYSB
Entrez Gene ID
493637
Gene Name
intestinal lysozyme
Gene Aliases
-
Chromosome Location
Chr.5: 44347290 - 44352877 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007805.1 NP_001007806.1 2 - 3 317 136
GenBank mRNA AY684065.1 - 2 - 3 317 136
BC102281.1 - 2 - 3 328 136

View Full Product Details