Assay Design



Gene Transcripts

Gene Symbol
PAG1
Entrez Gene ID
281964
Gene Name
pregnancy-associated glycoprotein 1
Gene Aliases
PAG1B
Chromosome Location
Chr.29: 38604865 - 38614193 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174411.2 NP_776836.1 1 - 2 66 106
GenBank mRNA BC134743.1 - 1 - 2 95 106
M73962.1 - 1 - 2 66 106

View Full Product Details