Assay Design



Gene Transcripts

Gene Symbol
BOLA
Entrez Gene ID
515712
Gene Name
MHC class I heavy chain
Gene Aliases
3349.1, BoLA-N, Bota, D18.5, LA-A11
Chromosome Location
Chr.23: 28720499 - 28724353 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038518.2 NP_001033607.1 5 - 6 1015 165
GenBank mRNA BC109586.1 - 5 - 6 1016 165

View Full Product Details