Assay Design



Gene Transcripts

Gene Symbol
BOLA
Entrez Gene ID
515712
Gene Name
MHC class I heavy chain
Gene Aliases
3349.1, BoLA-N, Bota, D18.5, LA-A11
Chromosome Location
Chr.23: 28720499 - 28724353 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038518.2 NP_001033607.1 6 - 7 1054 141
GenBank mRNA AB008627.1 - 6 - 7 979 141
BC109586.1 - 6 - 7 1055 141
BC149733.1 - - 141

View Full Product Details