Assay Design



Gene Transcripts

Gene Symbol
ZNF75D
Entrez Gene ID
614260
Gene Name
zinc finger protein 75D
Gene Aliases
ZNF75
Chromosome Location
Chr.X: 19043468 - 19061106 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078043.1 NP_001071511.1 2 - 3 743 76
GenBank mRNA BC122656.1 - 2 - 3 743 76

View Full Product Details