Assay Design



Gene Transcripts

Gene Symbol
HNMT
Entrez Gene ID
613413
Gene Name
histamine N-methyltransferase
Gene Aliases
-
Chromosome Location
Chr.2: 59138783 - 59180279 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035434.1 NP_001030511.1 6 - 6 1246 146
GenBank mRNA BC114060.1 - 6 - 6 1308 146
BT021490.1 - 6 - 6 1246 146

View Full Product Details