Assay Design



Gene Transcripts

Gene Symbol
FAHD1
Entrez Gene ID
509273
Gene Name
fumarylacetoacetate hydrolase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.25: 1392519 - 1394143 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075400.1 NP_001068868.1 1 - 1 1556 146
GenBank mRNA BC113240.1 - 1 - 1 1556 146

View Full Product Details