Assay Design



Gene Transcripts

Gene Symbol
THAP11
Entrez Gene ID
539845
Gene Name
THAP domain containing 11
Gene Aliases
-
Chromosome Location
Chr.18: 35349944 - 35351618 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001104994.1 NP_001098464.1 1 - 1 1448 129
GenBank mRNA BC142469.1 - 1 - 1 1448 129

View Full Product Details