Assay Design



Gene Transcripts

Gene Symbol
GLYATL3
Entrez Gene ID
787783
Gene Name
glycine-N-acyltransferase like 3
Gene Aliases
-
Chromosome Location
Chr.23: 22288176 - 22308391 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001145061.1 NP_001138533.1 2 - 3 213 95
GenBank mRNA DT841858.1 - 2 - 3 214 95

View Full Product Details