Assay Design



Gene Transcripts

Gene Symbol
TMX4
Entrez Gene ID
618015
Gene Name
thioredoxin related transmembrane protein 4
Gene Aliases
TXNDC13
Chromosome Location
Chr.13: 680566 - 739284 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099170.1 NP_001092640.1 4 - 5 560 63
GenBank mRNA BC146095.1 - 4 - 5 560 63

View Full Product Details