Assay Design



Gene Transcripts

Gene Symbol
VGLL2
Entrez Gene ID
541260
Gene Name
vestigial like family member 2
Gene Aliases
-
Chromosome Location
Chr.9: 33499510 - 33507296 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078032.1 NP_001071500.1 2 - 3 586 85
GenBank mRNA BC126731.1 - 2 - 3 586 85

View Full Product Details