Assay Design



Gene Transcripts

Gene Symbol
TPRN
Entrez Gene ID
100125320
Gene Name
taperin
Gene Aliases
-
Chromosome Location
Chr.11: 106213509 - 106220998 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103350.2 NP_001096820.2 2 - 3 2095 109
GenBank mRNA BC149384.1 - 2 - 3 1390 109

View Full Product Details