Assay Design



Gene Transcripts

Gene Symbol
NKPD1
Entrez Gene ID
789234
Gene Name
NTPase KAP family P-loop domain containing 1
Gene Aliases
-
Chromosome Location
Chr.18: 52806783 - 52817819 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163482.2 NP_001156954.2 1 - 2 78 79
GenBank mRNA EV686643.1 - 1 - 2 80 79

View Full Product Details