Assay Design



Gene Transcripts

Gene Symbol
MARCHF11
Entrez Gene ID
781742
Gene Name
membrane associated ring-CH-type finger 11
Gene Aliases
MARCH11
Chromosome Location
Chr.20: 56953217 - 57071178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205420.1 NP_001192349.1 2 - 3 691 83

View Full Product Details