Assay Design



Gene Transcripts

Gene Symbol
PDZRN4
Entrez Gene ID
511963
Gene Name
PDZ domain containing ring finger 4
Gene Aliases
-
Chromosome Location
Chr.5: 39200826 - 39632049 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205492.1 NP_001192421.1 9 - 10 1584 102

View Full Product Details