Assay Design



Gene Transcripts

Gene Symbol
KLK15
Entrez Gene ID
100140565
Gene Name
kallikrein related peptidase 15
Gene Aliases
KLNE
Chromosome Location
Chr.18: 56887389 - 56895414 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205495.1 NP_001192424.1 4 - 5 618 71

View Full Product Details