Assay Design



Gene Transcripts

Gene Symbol
KLHL23
Entrez Gene ID
100140861
Gene Name
kelch like family member 23
Gene Aliases
-
Chromosome Location
Chr.2: 26570154 - 26589955 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205507.1 NP_001192436.1 2 - 3 1425 75

View Full Product Details