Assay Design



Gene Transcripts

Gene Symbol
UROD
Entrez Gene ID
504914
Gene Name
uroporphyrinogen decarboxylase
Gene Aliases
-
Chromosome Location
Chr.3: 101034183 - 101037522 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001205558.1 NP_001192487.1 4 - 5 326 74

View Full Product Details